Learn More
Honeywell™ Dispensing System and Dip Tube
Shop All Honeywell Fluka Products
Click to view available options
For Use With (Equipment):
18 L Drum without Dip Tube
200 L Drum without Dip Tube
45 L Drum without Dip Tube
Capacity (Metric):
18 L
45 L
200 L
Unit Size:
1 pack
Specifications
Specifications
| Product Type | Returnable Container Accessory |
| Capacity (Metric) | 45 L |
| For Use With (Equipment) | 45 L Drum without Dip Tube |
| Quantity | 1 |
| Weight (Metric) | 1.22 Kg |