Learn More
BRAND™ Dispensing Cylinder with Valve Block
For use with Titrette™ Bottletop Burette. Accommodates all serial number ranges.
Brand: BRAND™ 707537
88.30 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with Titrette™ Bottletop Burette
- Fits all serial number ranges
- *For 50mL instruments with serial numbers beginning with 01-12 AND letters A-J, must order both Dispensing cylinder and Titrating tube as a pair.
Specifications
Dispensing Cylinder | |
Valve head for titrette | |
50 mL Titrette bottletop burette |
BRAND Titrette Bottle-top burette - English.PDF
File size: 1.3 MB
Product Content Correction
Product Title
BRAND™ Dispensing Cylinder with Valve Block > Dispensing cylinder; For 50mL Titrette bottletop burette