Learn More
Bode Chemie™ Dismofix™ G Gloss Finish Cleaner
For the maintenance cleaning of all washable surfaces and coated floors.
Brand: Bode Chemie™ 9802003
This item is not returnable.
View return policy
Description
- Dries without leaving streaks
- Low foaming
- For streak-free cleaning of sensitive surfaces such as glass, plastics and ceramics
- For automated cleaning of all treated floors with cleaning machines
- For the cleaning of conductive flooring
Specifications
Dismofix G Gloss Finish Cleaner | |
7 (at 20°C) | |
Cleaning |
Liquid | |
Yellow | |
5 L |
Product Content Correction
Product Title
Bode Chemie™ Dismofix™ G Gloss Finish Cleaner > Dismofix G Gloss Finish Cleaner