Learn More
KGW-Isotherm Dish-Shaped Dewer Flasks, Aluminum Cover
Reliable and economical Dewar flasks according to DIN EN ISO 16496. KGW-Isotherm Dish-Shaped Dewer Flasks, Aluminum Cover pairs a bowl shaped glass flask in a protective casing made of stucco aluminum for easy handling. Lid sold separately.
€ 166.42 - € 204.58
Specifications
Color | Silver |
---|---|
Base Material | Aluminum |
Material | Borosilicate Glass |
Style | Fully silvered |
For Use With (Application) | Tempering, Cooling, Warming, Stirring |
Product Code | Brand | Model | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | ||
10323241
|
KGW-Isotherm
1191 |
SCH 6 CAL | 80 mL | 80-90 mm | 75 mm |
€ 166.42
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
10179600
|
KGW-Isotherm
1192 |
SCH 9 CAL | 120 mL | 95-110 mm | 90 mm |
€ 204.58
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
||
Description
- Made of borosilicate glass according to ISO 3585
- Structured aluminum cover: antimagnetic and stainless
- Fully silvered
- Applications:
- tempering of round bottom flasks
- cooling of small samples
- suitable as a hot or cold bath
- suitable on magnetic stirrer
- Chemical characteristics:
- Hydrolytic resistance: according to ISO 719 (98°C)
- Hydrolytic resistance: according to ISO 720 (121°C)
- Acid resistance: according to ISO 1776
- Alkaline resistance: according to ISO 695-A2
- Temperature range:
- Glass refill from -200°C to + 150°C, for a short time 200°C
- Casing with mounting rubber and Latizell buffer from -10°C to + 60°C
Specifications
Silver | |
Borosilicate Glass | |
Tempering, Cooling, Warming, Stirring |
Aluminum | |
Fully silvered |