Learn More
B Braun™ Discofix™ C 3-Way Stopcocks
Click to view available options
Color:
Blue
Red
Unit Size:
Pack of 50
Description
- Resistant to the effects of pharmaceuticals with a previously unattained degree of transparency
- Pressure resistant up to 2 bar
- Free of latex, PVC and DEHP
- Resistance against stress cracking resulting from exposure to infusion solutions, drugs and alcohol-based disinfectants
- Protection against the danger of unnoticed cracks, air infusion and leakage
- Fast connection with rotating adapter
- Reliable, secure system connection with rotation of the connected stopcock unit on its own axis
- Light snap at every 45° turn of the valve
- Allows convenient access for injections and aspirations by rotating adapter
- Lets you feel the proper setting by tactile response
- Applications include parenteral nutrition therapy, anesthesia with IV, chemotherapy with cytostatic agents, immunotherapy and neurology
Specifications
Specifications
| Color | Red |
| For Use With (Application) | Infusion therapy and monitoring |
| Latex Free | Yes |
| Product Type | Stopcock |
| Max. Pressure | 2 bar |
| Quantity | 100/Pk. |