Learn More
Esculape Laboratoire™ Diphoterine Station
Neutralises and decontaminates chemical burns on hands, face or body caused by strong chemicals.
Brand: Esculape Laboratoire™ 8205206
This item is not returnable.
View return policy
Description
- Neutralises chemicals burns from acids, bases and solvents
- Physiological pH returns in seconds
- Ophthalmic solution rinses and rehydrates the eye
- Durable cabinet with lid protects contents from dust and splashes
- Wall-mounting
- Two year shelf life
Specifications
Chemical Splashes and Burns | |
2 x 500 mL Diphoterine eyewash, 1 x 200 mL ophthalmic eyewash solution, 1 x 200 mL Diphoterine spray |
Product Content Correction
Product Title
Esculape Laboratoire™ Diphoterine Station > Chemical splashes and burns