Learn More
Thermo Scientific™ Dionex™ Specially Formulated PTFE Tubing
Shop All Thermo Scientific Products
Click to view available options
Description:
250 and 350 um ID
250 um ID; Low pressure
350 um ID; Low pressure
For Use With (Equipment):
Dionex UltiMate 3000 RSLC Nano System
Fused Silica Capillary Tubing
Pakningsstørrelse:
Pack of 2
Pack of 5
Beskrivelse
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC).
Tekniske data
Tekniske data
| Description | 250 um ID; Low pressure |
| For Use With (Equipment) | Dionex UltiMate 3000 RSLC Nano System |
| Type | Capillary Tubing |
| Product Line | nanoLC |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion