Learn More
Thermo Scientific™ Dionex™ PEEK Sleeves
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Connections with Fused Silica Tubing
Connections with Microtight Union
Diameter (Metric) Outer:
30 μm
280 μm
360 μm
Unit Size:
Pack of 10
Pack of 5
Description
Thermo Scientific™ Dionex PEEK Sleeves are precision cut and polished.
Spécification
Spécification
| Description | For fused silica tubing; 280 um OD |
| For Use With (Equipment) | Connections with Fused Silica Tubing |
| Type | Tubing Sleeve |
| Diameter (Metric) Outer | 280 μm |
| Product Line | nanoLC |