Learn More
OHAUS™ Dimpled Mat
OHAUS™ Dimpled Mat designed to securely hold tubes in place.
Brand: OHAUS™ 30400140
36.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
- Platforms can increase the amount of usable space for samples
- In some cases can be stacked to increase throughput and double sample capacity
- Tube racks and dimpled mats securely hold tubes in place
Specifications
Platform Mat | |
12.75 in. | |
32.3 x 25.4 cm | |
32.40 cm | |
25.40 cm | |
Rocker |
PVC | |
32.40 cm | |
12.75 in. | |
10.00 in. | |
Dimpled Mat For Rocker SHRK04DG |
Product Content Correction
Product Title
OHAUS™ Dimpled Mat > Dimensions (L x W): 32.3 x 25.4 cm