missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DimethylFormamide, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Amber glass bottle
HDPE plastic bottle
Plastic Coated Glass bottle
Plastic container
Description
DimethylFormamide, Certified AR for Analysis

Specifications
Specifications
Melting Point | -61°C |
Color | Colorless |
Boiling Point | 153°C |
Packaging | Plastic container |
Quantity | 25 L |
Formula Weight | 73.09g/mol |
Vapor Pressure | 4.9mbar at 20°C |
Physical Form | Liquid |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2527792 | Certificate of Analysis | 24/02/2025 | D/3841/25,D/3841/24,D/3841/27,D/3841/15,D/3841/PC15,D/3841/15X,D/3841/08 |
Safety and Handling



Product Identifier
- Dimethylformamide
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DimethylFormamide, Certified AR for Analysis, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction