missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Fluid Thioglycollate Medium
Click to view available options
Quantity:
10,000 g
2000 g
500 g
Packaging Type:
Bottle
Pail
Media Type:
Fluid Thioglycollate Medium
Unit Size:
10kg
2kg
500g
Description
- Formulation: Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
Certifications
AOAC, BAM, BS12, COMPF, EP, USDA, USP
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, BS12, COMPF, EP, USDA, USP |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Fluid Thioglycollate Medium |
| Product Type | Microbiology Media |
| Formulation | Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L |
| For Use With (Application) | Used for sterility testing |
| Packaging Type | Pail |
| Quantity | 10,000 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction