missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Fluid Thioglycollate Medium

Product Code. 11742834
Click to view available options
Quantity:
10,000 g
2000 g
500 g
Packaging Type:
Bottle
Pail
Media Type:
Fluid Thioglycollate Medium
Unit Size:
10kg
2kg
500g
This item is not returnable. View return policy

Product Code. 11742834

Brand: BD 225620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For sterility testing of biologics and cultivation of anaerobes, aerobes and microaerophiles

  • Formulation: Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L

Certifications

AOAC, BAM, BS12, COMPF, EP, USDA, USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, BS12, COMPF, EP, USDA, USP
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Fluid Thioglycollate Medium
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
For Use With (Application) Used for sterility testing
Packaging Type Pail
Quantity 2000 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.