missing translation for 'onlineSavingsMsg'
Learn More

Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

€ 63.62 - € 300.50

Chemical Identifiers

CAS 96-10-6, 108-88-3
Molecular Formula C4H10AlCl
Molecular Weight (g/mol) 120.56
MDL Number MFCD00000459
InChI Key YNLAOSYQHBDIKW-UHFFFAOYSA-M
Synonym diethylaluminum chloride, aluminum, chlorodiethyl, diethylaluminium chloride, deac, chlorodiethylaluminum, diethylchloroaluminum, diethylaluminum monochloride, deak, aluminum diethyl monochloride, aluminum, dichlorotetraethyldi
PubChem CID 7277
IUPAC Name chloro(diethyl)alumane
SMILES CC[Al](Cl)CC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10493083
SDS View Documents
Thermo Scientific Acros
381151000
100 mL AcroSeal™ Glass Bottle
€ 63.62
100mL
Please to purchase this item. Need a web account? Register with us today!
10576943
SDS View Documents
Thermo Scientific Acros
381158000
800 mL AcroSeal™ Glass Bottle
€ 300.50
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.