missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Diethanolamine (DEA) Buffer
For use with concentrated Tropix™ substrates for detection of alkaline phosphatase.
Brand: Invitrogen™ T2027
This item is not returnable.
View return policy
Description
- DEA buffer provide favorable reaction kinetics and greater sensitivity compared to most buffer systems
- Purity: 99% pure
- M.W.: 105.1g/mol
- Density: 1.097g/mL
- Suggested working concentration: 0.1M DEA (pH 10) with 1mM MgCl2
Specifications
Immunoassays, AP substrate detection | |
Store at room temperature | |
Diethanolamine (DEA) Buffer | |
120 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction