missing translation for 'onlineSavingsMsg'
Learn More

Smeg™ Deterliquid SP2

Liquid alkaline detergent for water analysis and pharmaceutics.

Brand:  Smeg™ DETERLQSP2


Product Code. 15874832

  • € 115.00 / 5 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Liquid alkaline detergent for water analysis and pharmaceutics.

  • Contains potassium hydroxide, sodium hydroxide
  • Phosphate-free
Specifications

Specifications

Can
Water Analysis and Pharmaceutics
1.32 gal.
5 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide,sodium hydroxide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Smeg™ Deterliquid SP2 > Deterliquid SP2; For Use With (Application): Water Analysis and Pharmaceutics; Form: Can; Volume (English): 1.32 gal.; Volume (Metric): 5 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.