missing translation for 'onlineSavingsMsg'
Learn More

Malathion-(diethyl-d10), Honeywell Fluka™

Product Code. 15987422
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY

Specifications

CAS 347841-48-9
Packaging Glass Bottle
Quantity 10 mg
Grade Analytical Standard
Density 1.23 g/cm3
Boiling Point 156°C
Flash Point 163°C
For Use With (Application) Acaricides and Insecticides
Physical Form Liquid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'