Learn More
Density Standards (20 °C), Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 20°C
€ 74.00 - € 85.05
Product Code | Brand | Density | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Density | Price | Quantity & Availability | |||||
13480489
|
Paragon Scientific
DEN20-01 |
0.6597 g/mL (20°C) |
€ 74.00
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13470489
|
Paragon Scientific
DEN20-02 |
0.7264 g/mL (20°C) |
€ 74.00
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13490489
|
Paragon Scientific
DEN20-03 |
0.7788 g/mL (20°C) |
€ 85.00
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13400499
|
Paragon Scientific
DEN20-04 |
0.8383 g/mL (20°C) |
€ 85.00
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13420499
|
Paragon Scientific
DEN20-05 |
0.8556 g/mL (20°C) |
€ 85.00
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13450499
|
Paragon Scientific
DEN20-06 |
0.8708 g/mL (20°C) |
€ 74.00
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13440499
|
Paragon Scientific
DEN20-07 |
0.9376 g/mL (20°C) |
€ 85.00
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13430499
|
Paragon Scientific
DEN20-08 |
0.9817 g/mL (20°C) |
€ 74.00
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13460499
|
Paragon Scientific
DEN20-09 |
1.0257 g/mL (20°C) |
€ 85.00
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13490499
|
Paragon Scientific
DEN20-10 |
1.0677 g/mL (20°C) |
€ 74.00
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13480499
|
Paragon Scientific
DEN20-11 |
1.1258 g/mL (20°C) |
€ 74.00
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13400509
|
Paragon Scientific
DEN20-12 |
1.1917 g/mL (20°C) |
€ 85.00
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13410509
|
Paragon Scientific
DEN20-13 |
1.2849 g/mL (20°C) |
€ 85.05
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13420509
|
Paragon Scientific
DEN20-14 |
1.6226 g/mL (20°C) |
€ 74.00
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in accordance with primary level ASTM D1480 methodology
- Ensures full compliance to ASTM and IP test method protocols.
- Supplied in tamper evident packaging (please refer to standard for shelf life).
- Manufactured in the United Kingdom and available from stock for immediate dispatch
Specifications
60 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- DEN20-01
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Reproductive toxicity Category 2
- Flammable liquid Category 2
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P273-Avoid release to the environment.
Nominal values quoted