Learn More
Eppendorf™ Delivery to Factory Gate
Click to view available options
For Use With (Application):
Delivery of >100, <180 kg Equipment
Delivery of >180 kg Equipment
Delivery of <100 kg Equipment
Unit Size:
Each
Description
- Delivery to the factory gate
- Delivery to first door at the end customer side
Specifications
Specifications
| Description | Delivery to factory gate (>180 kg) |
| Service Offering | Factory Gate Delivery |
| For Use With (Application) | Delivery of >180 kg Equipment |