Learn More
Ohaus™ Defender 3000 Storage Platform Scale
Shop All Ohaus Products
Click to view available options
Model:
D31XW300VX-EU
Unit Size:
Each
Specifications
Specifications
| Construction | Stainless Steel with IP66 Stainless Steel platform |
| Capacity | 300 kg |
| Display Type | LCD |
| Readability | 50 g |
| Model | D31XW300VX-EU |
| Protection Class | IP67 |
| Temperature (Metric) Operating | -10°C to +40°C |
| Type | Defender™ Storage Platform Scale |