Learn More
Helios Italquartz Deep and Shallow Crucibles, Translucent
Product Code. p-7156860
Click to view available options
Capacity (Metric):
12 cc
20 cc
5 cc
Diameter (Metric) Outer:
29 mm
32 mm
38 mm
Height (Metric):
19 mm
32 mm
38 mm
Unit Size:
Each
Product Code. 15828044
Brand: Helios Italquartz 63F00502
Only null left
Add to basket
Add to basket
Deep and shallow crucibles constructed of translucent, durable, resistant, and chemically inert quartz glass.
  • Translucent
  • Deep and shallow forms available
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Color Translucent
Material Quartz
Capacity (Metric) 5 cc
Diameter (Metric) Outer 29 mm
Height (Metric) 19 mm
Product Type Crucible