Learn More
ValidR BV Decontamination Biosafety Cabinet
Decontamination of your incubator, biosafety cabinet, or freezer.
Brand:  ValidR BV BSC_DECONTAMINATION_2
Product Code. 30041501
  • € 1375.00 / Each
Explore more special offers
Description
Description
  • Equipment is disinfected using a hydrogen peroxide solution. The effectiveness of hydrogen peroxide ensures proper decontamination of your incubator, biosafety cabinet, or freezer.
  • We perform these services on-site, ideal for smaller equipment with at least 0.5 m³ of workspace. After decontamination, you will receive a detailed certificate.
  • General Terms and Conditions of Delivery ValidR B.V apply.
Specifications
Specifications
Labconco PSM and BSC Cabinets
4 to 8 hr.
Installation, Connection, and Commissioning Service