Learn More
ValidR BV Decontamination Biosafety Cabinet
Decontamination of your incubator, biosafety cabinet, or freezer.
Brand:  ValidR BV BSC_DECONTAMINATION_3
Product Code. 30041488
  • € 2200.00 / Each
Explore more special offers
Description
Description
  • Equipment is disinfected using a hydrogen peroxide solution. The effectiveness of hydrogen peroxide ensures proper decontamination of your incubator, biosafety cabinet, or freezer.
  • We perform these services on-site, ideal for smaller equipment with at least 0.5 m³ of workspace. After decontamination, you will receive a detailed certificate.
  • General Terms and Conditions of Delivery ValidR B.V apply.
Specifications
Specifications
Labconco PSM and BSC Cabinets
1.5 Days
Installation, Connection, and Commissioning Service
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers