Learn More
ValidR BV Decontamination Biosafety Cabinet
Decontamination of your incubator, biosafety cabinet, or freezer.
Brand: ValidR BV BSC_DECONTAMINATION_1
This item is not returnable.
View return policy
Description
- Equipment is disinfected using a hydrogen peroxide solution. The effectiveness of hydrogen peroxide ensures proper decontamination of your incubator, biosafety cabinet, or freezer.
- We perform these services on-site, ideal for smaller equipment with at least 0.5 m³ of workspace. After decontamination, you will receive a detailed certificate.
- General Terms and Conditions of Delivery ValidR B.V apply.
Specifications
Labconco PSM and BSC Cabinets | |
NEN EN 12469 | |
2 to 4 hr. | |
Installation, Connection, and Commissioning Service |