Learn More
Decitabine, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Decitabine (NSC 127716) is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. Decitabine incorporates into DNA in place of cytosine can covalently trap DNA methyltransferase to DNA causing irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis. Decitabine has potent anticancer activity.
Specifications
Specifications
| Chemical Name or Material | Decitabine |
| CAS | 2353-33-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C8H12N4O4 |
| Quantity | 10 mg |
| Synonym | 5-Aza-2'-deoxycytidine 5-AZA-CdR NSC 127716 |
| Solubility Information | DMSO : ≥ 50 mg/mL (219.10 mM) ∣H2O : 20 mg/mL (87.64 mM; Need ultrasonic) |
| SMILES | O=C1N=C(N)N=CN1[C@H](O[C@@H]2CO)C[C@@H]2O |
| Molecular Weight (g/mol) | 228.21 |
| Show More |
Research purposes only