Learn More
Trajan Scientific and Medical™ Deactivated (Methyl) Fused Silica Tubing
Trajan methyl deactivated fused silica capillary tubing is an essential ingredient in maintaining a high performance chromatography system.
Brand:  Trajan Scientific and Medical™ 0624434
 View more versions of this product
Product Code. 15208803
Quantity Price
1 € 176.00 / 10m
6 € 145.02 / 10m
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Description
Trajan methyl deactivated fused silica capillary tubing is an essential ingredient in maintaining a high performance chromatography system.
  • Chemically inert and thermally stable up to 380°C
  • Suitable for organic and aqueous solvents
  • Ideal for biotechnology applications
Specifications
Specifications
Organic and Aqueous Solvents
1/Pk.
363 μm
Thermally stable up to 380°C
Tubing
250 μm
10 m