missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DDR1-IN-1 dihydrochloride
Selective DDR1 inhibitor
€ 304.00 - € 1215.00
Specifications
Purity | >98% |
---|---|
CAS | 1780303-76-5 |
Product Type | DDR1-IN-1 dihydrochloride |
Molecular Weight (g/mol) | 625.51 |
Inhibitors | DDR1-IN-1 dihydrochloride |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18711558
|
Tocris Bioscience™
5077/10 |
10 mg |
€ 304.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18771779
|
Tocris Bioscience™
5077/50 |
50 mg |
€ 1215.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective discoidin domain receptor 1 (DDR1) tyrosine kinase inhibitor (IC50 values are 105 and 413 nM for DDR1 and DDR2 respectively). Inhibits integrin-induced DDR1 autophosphorylation in an osteosarcoma cell line.Specifications
>98% | |
DDR1-IN-1 dihydrochloride | |
DDR1-IN-1 dihydrochloride |
1780303-76-5 | |
625.51 |
For Research Use Only