Learn More
Thermo Scientific Chemicals D(+)-Sucrose, 99.7%, for biochemistry Product Code.: 15224664
Thermo Scientific Chemicals D(+)-Sucrose, 99.7%, for biochemistry
Product Code. 15224664
25 kg, Fiber Drum
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Plastic bottle
Plastic drum
Product Code. 15224664
Brand: Thermo Scientific Chemicals 177140250
Request Bulk or Custom Format
D(+)-Sucrose, CAS # 57-50-1, is a disaccharide used in a variety of biochemical applications such as protein purification. It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • D(+)-Sucrose is a disaccharide carbohydrate
  • It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose
Applications
  • D(+)-Sucrose can be used for protein and nucleic acid purification
  • Its fatty acid esters have laboratory research applications in transdermal systems
  • It is also used for sucrose-gradient centrifugation, which may be employed for the selective isolation of Nocardia species from soil
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Specifications
Color Colorless
pH 6.5 to 7.5 (10% soln.)
Loss on Drying 0.1 % max.
Infrared Spectrum Authentic
Assay Percent Range 0.997
Packaging Fiber Drum
Quantity 25 kg
Specific Rotation (20°C, 589nm) (c=26, H2O) > +65.9°
Solubility Information 0.0007 % max. Insoluble in Water
Formula Weight 342.29
Percent Purity ≥99.7% (HPLC)
Grade Biochemical
Physical Form Liquid
Chemical Name or Material D(+)-Sucrose
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463305 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog Number177140250,177140010,177142500,177140050
Lot NumberA0453886 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number177140250,177140010,177142500,177140050
Lot NumberA0453885 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number177140250,177140010,177142500,177140050
Lot NumberA0447978 Certificate TypeCertificate of Analysis Date10/04/2023 Catalog Number177140250,177140010,177142500,177140050
Lot NumberA0420137 Certificate TypeCertificate of Analysis Date23/12/2022 Catalog Number177140250,177140010,177142500,177140050
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals D(+)-Sucrose, 99.7%, for biochemistry >