missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals β-D-Galactose Pentaacetate ≥99%
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Color | White to Gray |
Infrared Spectrum | Authentic |
Assay Percent Range | 99+% |
Packaging | Plastic bottle |
Quantity | 100 g |
Beilstein | 31, 305 |
Specific Rotation Condition | +25° (20°C c=10,CHC3,on dry ba) |
Specific Rotation | +24° to +26° (20°C, 589nm) (c=2, CHCl3) |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0452853 | Certificate of Analysis | 16/05/2023 | 158611000,158610250 |
A0441101 | Certificate of Analysis | 28/04/2022 | 158611000,158610250 |
A0436417 | Certificate of Analysis | 19/11/2021 | 158611000,158610250 |
A0403344 | Certificate of Analysis | 10/06/2021 | 158611000,158610250 |
Safety and Handling
Product Identifier
- ß-D-Galactose pentaacetate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals β-D-Galactose Pentaacetate ≥99% >
Spot an opportunity for improvement?Share a Content Correction