Learn More
R&D Systems™ Cyno Monkey BAFF/TNFSF13B (XP_005586281) VersaClone cDNA
Brand: R&D Systems™ RDC2851
Specifications
XP_005586281 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
10673 | |
Cynomolgus Monkey |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Product Title
R&D Systems™ Cyno Monkey BAFF/TNFSF13B (XP_005586281) VersaClone cDNA > 10 ug