Learn More
Kartell Cylindrical Test Tube with Screw Cap, Polystyrene
Cylindrical test tubes equipped with a tight screw cap.
Brand: Kartell 8820000
This item is not returnable.
View return policy
Description
- Test tubes are intended to contain blood and biological liquids to perform laboratory tests
Specifications
Polypropylene | |
100 mm | |
Cylindrical | |
Non-sterile | |
No | |
Test Tube |
10 mL | |
Polystyrene | |
Single-use | |
Not Autoclavable | |
Screw Cap |