Learn More
Fisherbrand™ PTFE-Coated Rare Earth Core Stir Bars, Cylindrical
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
12 mm
20 mm
30 mm
50 mm
Diameter (Metric):
4.5 mm
6 mm
8 mm
missing translation for 'unitSize'
Pack of 10
Pack of 5
Description
- Use rare earth magnets for significantly increased magnetic strength compared to those using AlNiCo magnets
- Samarium-cobalt core is resistant to demagnetization
- Identified by a chemically inert, carbon black spot
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Samarium-Cobalt magnet core |
| For Use With (Equipment) | Containers & Vessels / Stirrer Hot Plates |
| Diameter (Metric) | 6 mm |
| Length (Metric) | 30 mm |
| Quantity | 2/Pk. |
| Shape | Cylinder |