Learn More
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Cone Size:
14/23 NS
29/32 NS
Includes:
Pressure Equalizing Tube
w/o Pressure Equalizing Tube
Packungsgröße:
Pack of 2
Description
- With NS socket and cone
- With scale (approx. volume/mL)
- With inscription label
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Material | Borosilicate Glass |
| Cone Size | 29/32 NS |
| Graduated | Yes |
| Includes | Pressure Equalizing Tube |
| Product Type | Dropping Funnel |