missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl Bromide, 99%, Thermo Scientific Chemicals

€ 15.77 - € 190.00

Chemical Identifiers

CAS 108-85-0
Molecular Formula C6H11Br
Molecular Weight (g/mol) 163.06
MDL Number MFCD00003819
InChI Key AQNQQHJNRPDOQV-UHFFFAOYSA-N
Synonym cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n
PubChem CID 7960
IUPAC Name bromocyclohexane
SMILES BrC1CCCCC1
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10081740
View Documents
Thermo Scientific Acros
159080050
5 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10397950
View Documents
Thermo Scientific Acros
159082500
250 mL Glass bottle
€ 55.28
250mL
Please to purchase this item. Need a web account? Register with us today!
10103450
View Documents
Thermo Scientific Acros
159080010
1 L Glass bottle
€ 106.53
1 L
Please to purchase this item. Need a web account? Register with us today!
10434025
View Documents
Thermo Scientific Acros
159080025
2.5 L Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-85-0
163.06
AQNQQHJNRPDOQV-UHFFFAOYSA-N
7960
BrC1CCCCC1
C6H11Br
MFCD00003819
cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n
bromocyclohexane

Specifications

108-85-0
1.3300g/mL
62°C
99%
C6H11Br
MFCD00003819
15, 2723
cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n
AQNQQHJNRPDOQV-UHFFFAOYSA-N
bromocyclohexane
7960
99%
Cyclohexyl bromide
Colorless to Yellow
166.0°C to 167.0°C
Authentic
C6H11Br
1.4940 to 1.4960
05, 24
1.33
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
BrC1CCCCC1
163.06
163.06
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
In use, may form flammable/explosive vapor-air mixture.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

missing translation for 'einecsNumber' : 203-622-2

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only