Learn More
B Braun™ Cutfix™ Disposable Scalpel Blades
Product Code. 10664511
Click to view available options
Size:
# 15
Unit Size:
Pack of 10
Product Code. 10664511
Brand: B Braun™ 5518032
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides precise cutting with double-sided blade holder. B Braun™ Cutfix™ Disposable Scalpel Blades are securely embedded in the solid plastic handle.
  • Each scalpel is sterile packed and ready for immediate use
  • Excellent cutting ability of the stainless steel blade
  • Ergonomic, solid plastic handle
  • Secure anchoring of the blade in the handle
Specifications
Blade Material Stainless Steel
Size # 15
Disposable Single-use
Sterility Sterile
Handle Material Plastic