Learn More
Invitrogen™ Custom RNA, 5' FLO
Custom RNA, 5' FLO
Brand: Invitrogen™ 10660067
This item is not returnable.
View return policy
Description
Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.
Specifications
Configuration Primer RNA 5' FLO, 20nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation | |
Configuration Primer RNA 5' FLO | |
20 nmol |
Product Content Correction
Product Title
Invitrogen™ Custom RNA, 5' FLO > 20 nmol