Learn More
Applied Biosystems™ Custom 5' Fluorescent Labeled/Unlabeled Pairs
Shop All Applied Biosystems Products
Click to view available options
Quantity:
10,000 pmol
300,000 pmol
80,000 pmol
Unit Size:
10000pmol
300000pmol
80000pmol
Description
A 5' fluorescent dye-labeled oligo and unlabeled complement are supplied in separate tubes. Choose among 6-FAM™, TET™, VIC™, HEX™, NED™, and PET™ as labels. Ideal for use in fragment analysis, gene expression studies, genotyping, and PCR. Oligos are desalted.
Specifications
Specifications
| Description | Custom 5' Labeled/Unlabeled Di-Repeat + Tail, 300000 pmol, 6-FAM™, TET™, VIC™, HEX™, NED™, PET™ 5' Reporter Dye, Choice of Delivered Scales, These Products can be used in a Variety of Applications |
| Product Type | Custom 5' Labeled or Unlabeled Di-Repeat + Tail |
| Content And Storage | -20°C |
| Quantity | 300,000 pmol |
| For Use With (Application) | Fragment Analysis, Gene Expression Studies, Genotyping, PCR |
| Form | Delivered either in Solution or Dried down |