Learn More
Curdione, MedChemExpress
MedChemExpress Curdione, one of the major sesquiterpene compounds from Rhizoma Curcumae, has been shown to exhibit multiple bioactive properties.
Brand: Medchem Express HY-N0353
This item is not returnable.
View return policy
Description
Curdione, one of the major sesquiterpene compounds from Rhizoma Curcumae, has been shown to exhibit multiple bioactive properties.Specifications
Curdione | |
White | |
C15H24O2 | |
(+)-Curdione | |
O=C1C[C@@H](C(C)C)C(C/C(C)=C/CC[C@@H]1C)=O | |
236.35 | |
Research |
13657-68-6 | |
Research | |
10 mg | |
DMSO : ≥ 100 mg/mL (423.10 mM) | |
236.35 | |
98.61% | |
Solid |
Research purposes only