Learn More
Thermo Scientific™ Culti-Loops™ Candida tropicalis ATCC™ 750™
Product Code. 11944082
Click to view available options
Quantity:
5 Loops/Pack
Unit Size:
Pack of 5
Product Code. 11944082
Brand: Thermo Scientific™ R4601240
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 750TM*
Description Candida tropicalis
Format Culti-Loops
Product Type Culti-Loops Quality Control Organism
Target Organism Class Fungi
Test Type Qualitative QC
Quantity 5 Loops/Pack
Strain Designations WDCM00112, 1909, ATCC 7349, ATCC4563,CBS 94, CCRC 20520, IFO 1070, IFO 1400, JCM 1541, NRRL Y-12699, NRRL Y-12968, NRRL Y-607]
Species Candida species
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.