Learn More
CleanSpace™ Technology CST Full Face Mask
A CST Full Face Mask is designed for use with the CleanSpace PRO Power Unit (CST1002) and CleanSpace ULTRA Power Unit (CST1012).
Brand: CleanSpace™ Technology CST1018
This item is not returnable.
View return policy
Description
- A CleanSpace CST Full Face Mask clips to the power unit and provides respiratory, face and eye protection.
- It is comprised of an outer visor, inner mask and a five-point harness to evenly support the mask and power unit and provide ease of movement.
- Compatible with the CleanSpace PRO Power Unit (CST1002) and CleanSpace ULTRA Power Unit (CST1012).
Specifications
500 full cycles | |
Clear | |
Full Face Mask | |
1 x Full Face Mask |
un3481 | |
None | |
Large |
Product Content Correction
Product Title
CleanSpace™ Technology CST Full Face Mask > Size: Large