Learn More
Thermo Scientific™ CryoMed™ Sensors
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
1.2/2.0 Cryo Tubes in CryoMed
4.0/5.0 Cryo Tubes in CryoMed
Unit Size:
Each
Description
Improve the functionality of Thermo Scientific™ Cryomed™ controlled-rate freezers with these CryoMed Sensors.
Features:
- Thermocouple sensors for cryo tubes, sheath straw and vial samples
- Includes male plug
Recommended for:
Controlled-rate freezing applications
Specifications
Specifications
| Description | Thermocouple Sensor |
| Type | Thermocouple Sensor |
| Capacity (Metric) | 5 mL |
| For Use With (Equipment) | 4.0/5.0 Cryo Tubes in CryoMed |
| Plug Type | Male |
| Holds | Cryo Tube, Sheath Straw, Vial |