Learn More
Crotonic acid, 98%, Thermo Scientific Chemicals
€ 45.63 - € 150.75
Chemical Identifiers
CAS | 107-93-7 |
---|---|
Molecular Formula | C4H6O2 |
Molecular Weight (g/mol) | 86.09 |
MDL Number | MFCD00002701 |
InChI Key | LDHQCZJRKDOVOX-NSCUHMNNSA-N |
Synonym | crotonic acid, e-but-2-enoic acid, but-2-enoic acid, trans-2-butenoic acid, trans-crotonic acid, 3-methylacrylic acid, e-2-butenoic acid, 2-butenoic acid, 2e-but-2-enoic acid, e-crotonic acid |
PubChem CID | 637090 |
ChEBI | CHEBI:41131 |
IUPAC Name | (E)-but-2-enoic acid |
SMILES | CC=CC(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10615972
|
Thermo Scientific Acros
150875000 |
500 g | Plastic bottle |
€ 45.63
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10414555
|
Thermo Scientific Acros
150870025 |
2.5 kg | Plastic bottle |
€ 150.75
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-93-7 | |
86.09 | |
LDHQCZJRKDOVOX-NSCUHMNNSA-N | |
637090 | |
(E)-but-2-enoic acid |
C4H6O2 | |
MFCD00002701 | |
crotonic acid, e-but-2-enoic acid, but-2-enoic acid, trans-2-butenoic acid, trans-crotonic acid, 3-methylacrylic acid, e-2-butenoic acid, 2-butenoic acid, 2e-but-2-enoic acid, e-crotonic acid | |
CHEBI:41131 | |
CC=CC(=O)O |
Specifications
107-93-7 | |
White to Yellow | |
88°C | |
97.5% min. (GC) | |
CH3CH=CHCO2H | |
02, IV, 1491 | |
crotonic acid, e-but-2-enoic acid, but-2-enoic acid, trans-2-butenoic acid, trans-crotonic acid, 3-methylacrylic acid, e-2-butenoic acid, 2-butenoic acid, 2e-but-2-enoic acid, e-crotonic acid | |
LDHQCZJRKDOVOX-NSCUHMNNSA-N | |
(E)-but-2-enoic acid | |
637090 | |
86.09 | |
Crystalline Flakes |
72.0°C | |
185.0°C | |
Authentic | |
C4H6O2 | |
MFCD00002701 | |
15, 2588 | |
Solubility in water: 94g/L (25°C). Other solubilities: soluble in ethanol,acetone and toluene | |
CC=CC(=O)O | |
86.09 | |
CHEBI:41131 | |
98% | |
Crotonic acid, 98% |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-533-9
missing translation for 'rtecsNumber' : GQ2900000
- Crotonic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only