Learn More
Microspec™ Cribbar Sample Bottles - Sterile Na2S2O3 Dosed
Cribbar Sample Bottles are sterile Na2S2O3 dosed
Brand:  Microspec™ Q9
 View more versions of this product
Product Code. 16635482
  • € 91.75 / Pack of 84
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cribbar Sample Bottles
  • Sterile Na2S2O3 Dosed
  • 18 mg/L
Specifications
Specifications
18 mg/L
Sterile
500 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers