Learn More
Abeba™ Crawler ALU 4500 Shoes
Aluminum toe cap provides lightweight toe protection. Sanitized™ treated.
Brand: Abeba™ 4500/45
This item is not returnable.
View return policy
Description
- CE EN ISO 20345:2011 S2 SRC
- Microfiber
- White/grey
- Aluminum toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3577)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
- Upper one piece
Specifications
CE EN ISO 20345:2011 S2 SRC | |
Microfiber | |
Slip-resistant | |
4500 |
White | |
45 | |
Polyurethane | |
Aluminum |