Learn More
Abeba™ Crawler ALU 34555 Shoes
Product Code. 15573844
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15573844
Brand: Abeba™ 34555/48
Only null left
Add to basket
Add to basket
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
  • CE EN ISO 20345:2011 SB A, E, FO SRC
  • Microfiber
  • White
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3579)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Breathable lining with Silver ions
  • Instep area adjustable, with elastic band
  • velcro fastening
  • Scratch-resistant coating in toe area
Specifications
Certifications/Compliance CE EN ISO 20345:2011 SB A, E, FO SRC
Color White
Material Microfiber
Size 48
Sole Features Slip-resistant
Sole Material Polyurethane
Style 34555
Toe Material Aluminum