Learn More
COX7C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11411-2-AP-150UL
 View more versions of this product
Product Code. 16818473
  • € 410.00 / 150µL
Description
This antibody is specific to COX7C.
Cytochrome c oxidase (COX), the terminal component of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. This component is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may function in the regulation and assembly of the complex.
Specifications
Specifications
COX7C
Polyclonal
Unconjugated
COX7C
COX7C
Rabbit
Antigen Affinity Chromatography
RUO
100188937, 12867, 1350
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P15954, P17665, P80432
COX7C
COX7C Fusion Protein Ag1989
150 μL
Primary
Human, Mouse, Rat