Specifications
Color | Yellow |
Material | Cowhide Leather |
Product Type | Gloves |
Size | 9 |
Thickness (Metric) | 1.3 mm |
Model | T09 |
Resistant To | Thermal, Cuts |
Product Content Correction
Product Title
Sperian™ Cowhide Leather Gloves >