Learn More
Sartorius™ Cotton Linters FN 5 Chromatography Paper
Shop All Sartorius Products
Click to view available options
Length (Metric):
580 mm
Unit Size:
Pack of 100
Specifications
Specifications
| Format | Sheet |
| Grade | FN 5 |
| Material | Cotton Linters |
| Width (Metric) | 600 mm |
| Length (Metric) | 580 mm |
| Technique | Chromatography |