Learn More
Corning™ Polypropylene PCR Tubes with Attached Cap
Shop All Corning Products
missing translation for 'orderingAttributeHoverText'
Color:
Clear
Closure Type:
Domed Cap
Flat Cap
Volume (Metric):
0.2 mL
0.5 mL
missing translation for 'unitSize'
Pack of 1000
Pack of 960
Description
Individual PCR tubes are made of thin wall polypropylene and designed for precise fit in heat blocks to optimize heat transfer. Tubes are RNase-/DNase-free, are autoclavable at 121°C, and withstand centrifugation to 10,000 x g.
Specifications
Specifications
| Certifications/Compliance | RNase, DNase-free |
| Closure Color | Clear |
| Color | Clear |
| Volume (Metric) | 0.2 mL |
| Quantity | 96/Pk. |
| Closure Material | Polypropylene |
| For Use With (Equipment) | Heat blocks for fast heat transfer |
| Shape | Conical bottom |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Show More |