Learn More
Corning™ Costar™ Netwell™ Reagent Trays, Non-Sterile
GREENER_CHOICE
Product Code. p-176999 Shop All Corning Products
Click to view available options
Color:
Black
White
Unit Size:
Pack of 200
Product Code. 10621272
Brand: Corning™ 3517
Only null left
Add to basket
Add to basket
Specially designed Netwell™ carriers with handles allow simultaneous processing of up to 12 samples per carrier
  • Specially designed Netwell carriers and handles allow simultaneous processing of up to 12 samples per carrier
  • Polystyrene reagent trays are available in white for colorimetric reaction contrast, or black for better visibility of tissue sections
  • Each carrier kit contains eight carriers and eight handles
TRUSTED_SUSTAINABILITY
Specifications
Type Tray
No. per Pack 25/Pk.
No. per Case 200/Cs.
Color Black