Learn More
Corning™ PTFE-Coated Magnetic Stir Bars
Shop All Corning Products
Click to view available options
Length (Metric):
25.4 mm
50.8 mm
76.2 mm
Diameter (Metric):
9.5 mm
19 mm
Length (English):
1 in.
2 in.
3 in.
Diameter (English):
0.37 in.
0.75 in.
Unit Size:
Each
Description
Corning magnetic stir bars are polytetrafluorethylene (PTFE)-coated for increased chemical resistance.
Specifications
Specifications
| Diameter (English) | 0.37 in. |
| For Use With (Equipment) | All PC-200 and 400 Series hot plates and stirrers |
| Length (English) | 1 in. |
| Diameter (Metric) | 9.5 mm |
| Length (Metric) | 25.4 mm |
| Material | Magnet |
| Quantity | 1 |